MAN326367510 Manchester Organics

(R)-1-Boc-2-benzyl-piperazine

  • MDL number MFCD07772094
  • Cas Number : 947684-78-8
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U22276
Availability details for
Quantity
  • Availability
  • Add to Cart