MAN326367511 Manchester Organics

(S)-1-Boc-2-benzyl-piperazine

  • MDL number MFCD03787923
  • Molecular Weight : 276.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/V22609

Safety Information

MSDS https://www.manchesterorganics.com/V22609
Availability details for
Quantity
  • Availability
  • Add to Cart