MAN326367518 Manchester Organics

(S)-1-Boc-3-benzyl-piperazine

  • MDL number MFCD07772102
  • Molecular Weight : 276.374

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C22286
Availability details for
Quantity
  • Availability
  • Add to Cart