MAN326367524 Manchester Organics

N-Benzoyl-2-(difluoromethyl)pyrrole

  • MDL number MFCD16140209
  • Cas Number : 163811-28-7
  • Molecular Weight : 221.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-benzoyl-2-(difluoromethyl)-1H-pyrrole
Inchi Code 1S/C12H9F2NO/c13-11(14)10-7-4-8-15(10)12(16)9-5-2-1-3-6-9/h1-8,11H
Inchi Key MTGNPQCKYUXXEZ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin GB
Product Link https://www.manchesterorganics.com/W14681
Availability details for
Quantity
  • Availability
  • Add to Cart