MAN326367527 Manchester Organics

2-(Boc-aminomethyl)pyrrolidine

  • MDL number MFCD06658352
  • Molecular Weight : 200.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J22363

Safety Information

MSDS https://www.manchesterorganics.com/J22363
Availability details for
Quantity
  • Availability
  • Add to Cart