MAN326367606 Manchester Organics

3,3,3-Trifluoro-2,2-dimethylpropionic acid methyl ester

  • MDL number MFCD18253569
  • Molecular Weight : 170.12966

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S23151

Safety Information

MSDS https://www.manchesterorganics.com/S23151
Availability details for
Quantity
  • Availability
  • Add to Cart