MAN326367628 Manchester Organics

4-Piperazine-piperidine trihydrochloride

  • MDL number MFCD11518985
  • Molecular Weight : 278.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S22370
Availability details for
Quantity
  • Availability
  • Add to Cart