MAN326367641 Manchester Organics

2,3,4,5-Tetrahydro-1H-3-benzazepine

  • MDL number MFCD04012620
  • Cas Number : 4424-20-8
  • Molecular Weight : 147.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/H12042
Availability details for
Quantity
  • Availability
  • Add to Cart