MAN326367643 Manchester Organics

1-Bromo-2,4,5-trichlorobenzene

  • MDL number MFCD00155012
  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/H22456

Safety Information

MSDS https://www.manchesterorganics.com/H22456
Availability details for
Quantity
  • Availability
  • Add to Cart