MAN326367652 Manchester Organics

(S)-tert-butyl pyrrolidin-2-ylmethylcarbamate

  • MDL number MFCD06796572
  • Cas Number : 141774-70-1
  • Molecular Weight : 200.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U22395
Availability details for
Quantity
  • Availability
  • Add to Cart