MAN326367669 Manchester Organics

tert-Butyl 1-benzylazetidin-3-ylcarbamate

  • MDL number MFCD09878667
  • Molecular Weight : 262.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P22581

Safety Information

MSDS https://www.manchesterorganics.com/P22581
Availability details for
Quantity
  • Availability
  • Add to Cart