MAN326367673 Manchester Organics

(2-Oxo-pyrrolidin-1-yl)-phenyl-acetic acid

  • MDL number MFCD11214950
  • Cas Number : 60729-82-0
  • Molecular Weight : 219.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-oxo-1-pyrrolidinyl)(phenyl)acetic acid
Inchi Code 1S/C12H13NO3/c14-10-7-4-8-13(10)11(12(15)16)9-5-2-1-3-6-9/h1-3,5-6,11H,4,7-8H2,(H,15,16)
Inchi Key JEZXSMOLJOXBRE-UHFFFAOYSA-N

Description

Purity 90%
Country of Origin GB
Product Link https://www.manchesterorganics.com/J12043
Availability details for
Quantity
  • Availability
  • Add to Cart