MAN326367681 Manchester Organics

Boc-D-Pro-OMe

  • MDL number MFCD00190828
  • Molecular Weight : 229.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/J22386

Safety Information

MSDS https://www.manchesterorganics.com/J22386
Availability details for
Quantity
  • Availability
  • Add to Cart