MAN326367691 Manchester Organics

1-N-Boc-4-azetidin-3-yl-piperazine

  • MDL number MFCD09264437
  • Molecular Weight : 241.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin GB
Product Link https://www.manchesterorganics.com/H22433
Availability details for
Quantity
  • Availability
  • Add to Cart