MAN326367698 Manchester Organics

(R)-1-(tert-butoxycarbonyl)piperidine-3-carboxylic acid

  • MDL number MFCD02179173
  • Molecular Weight : 229.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/S22629

Safety Information

MSDS https://www.manchesterorganics.com/S22629
Availability details for
Quantity
  • Availability
  • Add to Cart