MAN326367699 Manchester Organics

3-Cyanopiperidine

  • MDL number MFCD01321091
  • Cas Number : 7492-88-8
  • Molecular Weight : 110.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/T22442
Availability details for
Quantity
  • Availability
  • Add to Cart