MAN326367707 Manchester Organics

4-Cbz-piperazine-2-carboxylate methyl ester

  • MDL number MFCD03001724
  • Molecular Weight : 278.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A22472

Safety Information

MSDS https://www.manchesterorganics.com/A22472
Availability details for
Quantity
  • Availability
  • Add to Cart