MAN326367708 Manchester Organics

1-Boc-3-formylpyrrolidine

  • MDL number MFCD03791257
  • Cas Number : 59379-02-1
  • Molecular Weight : 199.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/B22450
Availability details for
Quantity
  • Availability
  • Add to Cart