MAN326367718 Manchester Organics

6-Methyl-2-piperidine carboxylic acid

  • MDL number MFCD00513436
  • Molecular Weight : 143.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/L22553

Safety Information

MSDS https://www.manchesterorganics.com/L22553
Availability details for
Quantity
  • Availability
  • Add to Cart