MAN326367766 Manchester Organics

Potassium benzyl N-[2-(trifluoroboranuidyl)ethyl]carbamate

  • MDL number MFCD10698612
  • Molecular Weight : 285.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/N22651

Safety Information

MSDS https://www.manchesterorganics.com/N22651
Availability details for
Quantity
  • Availability
  • Add to Cart