MAN326367771 Manchester Organics

3-Bromo-2-fluoro-6-(trifluoromethyl)benzaldehyde

  • MDL number MFCD11520664
  • Cas Number : 909186-28-3
  • Molecular Weight : 271.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/U22514
Availability details for
Quantity
  • Availability
  • Add to Cart