MAN326367807 Manchester Organics

1,3-Dimethyl-1H-pyrazol-5-ol

  • MDL number MFCD00129673
  • Molecular Weight : 112.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K22623

Safety Information

MSDS https://www.manchesterorganics.com/K22623
Availability details for
Quantity
  • Availability
  • Add to Cart