MAN326367812 Manchester Organics

(3-(1H-Pyrazol-1-yl)phenyl)methanamine

  • MDL number MFCD03449005
  • Molecular Weight : 173.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/R22724

Safety Information

MSDS https://www.manchesterorganics.com/R22724
Availability details for
Quantity
  • Availability
  • Add to Cart