MAN326367852 Manchester Organics

4-Bromo-pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD09839150
  • Molecular Weight : 230.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/B22638

Safety Information

MSDS https://www.manchesterorganics.com/B22638
Availability details for
Quantity
  • Availability
  • Add to Cart