MAN326368005 Manchester Organics

5-Fluoro-6-chlorooxindole

  • MDL number MFCD09839325
  • Molecular Weight : 185.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/K22813

Safety Information

MSDS https://www.manchesterorganics.com/K22813
Availability details for
Quantity
  • Availability
  • Add to Cart