MAN326368007 Manchester Organics

2,3-Dimethyl-5-fluoroindole

  • MDL number MFCD01158420
  • Cas Number : 526-47-6
  • Molecular Weight : 163.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/M22744
Availability details for
Quantity
  • Availability
  • Add to Cart