MAN326368016 Manchester Organics

(2R,4S)-4-Fluoropyrrolidine-2-carboxylic acid

  • MDL number MFCD07366527
  • Molecular Weight : 133.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/P22485

Safety Information

MSDS https://www.manchesterorganics.com/P22485
Availability details for
Quantity
  • Availability
  • Add to Cart