MAN326368077 Manchester Organics

2-Amino-3-fluoro-4-(trifluoromethyl)pyridine

  • MDL number MFCD16606170
  • Cas Number : 1227514-27-3
  • Molecular Weight : 180.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/W12054
Availability details for
Quantity
  • Availability
  • Add to Cart