MAN326368093 Manchester Organics

5,6-Dihydroxyindole

  • MDL number MFCD00798933
  • Cas Number : 3131-52-0
  • Molecular Weight : 149.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y22825
Availability details for
Quantity
  • Availability
  • Add to Cart