MAN326368099 Manchester Organics

5-Bromo-1H-Indole-3-Carboxylic Acid

  • MDL number MFCD05664007
  • Molecular Weight : 240.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 94
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E22831
Availability details for
Quantity
  • Availability
  • Add to Cart