MAN326368101 Manchester Organics

3-Bromo-2-(difluoromethyl)pyridine

  • MDL number MFCD16875638
  • Cas Number : 1211520-77-2
  • Molecular Weight : 208.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/V15367
Availability details for
Quantity
  • Availability
  • Add to Cart