MAN326368116 Manchester Organics

1-Boc-3-(cyanomethyl)azetidine

  • MDL number MFCD11035913
  • Molecular Weight : 196.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A22353

Safety Information

MSDS https://www.manchesterorganics.com/A22353
Availability details for
Quantity
  • Availability
  • Add to Cart