MAN326368124 Manchester Organics

5-Chlorogramine

  • MDL number MFCD01074497
  • Molecular Weight : 208.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C22900

Safety Information

MSDS https://www.manchesterorganics.com/C22900
Availability details for
Quantity
  • Availability
  • Add to Cart