MAN326368128 Manchester Organics

5-Chloroindole-3-carboxylic acid

  • MDL number MFCD03410308
  • Cas Number : 10406-05-0
  • Molecular Weight : 195.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/F22857
Availability details for
Quantity
  • Availability
  • Add to Cart