MAN326368167 Manchester Organics

6-Methylindole-3-carboxaldehyde

  • MDL number MFCD00049347
  • Molecular Weight : 159.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/X23344

Safety Information

MSDS https://www.manchesterorganics.com/X23344
Availability details for
Quantity
  • Availability
  • Add to Cart