MAN326368186 Manchester Organics

2,3,3-Trimethyl-5-methoxy-3H-indole

  • MDL number MFCD04972105
  • Molecular Weight : 189.254

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T22914
Availability details for
Quantity
  • Availability
  • Add to Cart