MAN326368194 Manchester Organics

4-Bromo-1-(n-hexyl)-benzene

  • MDL number MFCD00061114
  • Molecular Weight : 241.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin GB
Product Link https://www.manchesterorganics.com/Z21667
Availability details for
Quantity
  • Availability
  • Add to Cart