MAN326368202 Manchester Organics

2-Hydroxy-3-methylpiperazine

  • MDL number MFCD01646120
  • Molecular Weight : 110.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/C22971

Safety Information

MSDS https://www.manchesterorganics.com/C22971
Availability details for
Quantity
  • Availability
  • Add to Cart