MAN326368209 Manchester Organics

3-Chloropyrazine-2-yl-methylamine hydrochloride

  • MDL number MFCD09910171
  • Cas Number : 939412-86-9
  • Molecular Weight : 180.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-2-pyrazinyl)methanamine hydrochloride
Inchi Code 1S/C5H6ClN3.ClH/c6-5-4(3-7)8-1-2-9-5;/h1-2H,3,7H2;1H
Inchi Key YYVVOYJKQZWKFS-UHFFFAOYSA-N

Description

Country of Origin GB
Product Link https://www.manchesterorganics.com/J22931
Availability details for
Quantity
  • Availability
  • Add to Cart