MAN326368275 Manchester Organics

1-Boc-amino-1,4-cyclohexanediamine

  • MDL number MFCD01076211
  • Molecular Weight : 214.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/E22405

Safety Information

MSDS https://www.manchesterorganics.com/E22405
Availability details for
Quantity
  • Availability
  • Add to Cart