MAN326368277 Manchester Organics

3-Fluoro-2-methylpyridine

  • MDL number MFCD08705710
  • Cas Number : 15931-15-4
  • Molecular Weight : 111.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/W22516
Availability details for
Quantity
  • Availability
  • Add to Cart