MAN326368280 Manchester Organics

BOC-2-methylamino-ethylcarbamate

  • MDL number MFCD06808585
  • Cas Number : 122734-32-1
  • Molecular Weight : 174.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin GB
Product Link https://www.manchesterorganics.com/H22337
Availability details for
Quantity
  • Availability
  • Add to Cart