MAN326368309 Manchester Organics

1-Boc-3-(Aminomethyl)azetidine hydrochloride

  • MDL number MFCD11101335
  • Molecular Weight : 222.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/T22559

Safety Information

MSDS https://www.manchesterorganics.com/T22559
Availability details for
Quantity
  • Availability
  • Add to Cart