MAN326368317 Manchester Organics

6-Amino-5-bromopyridine-2-carboxylic acid methyl ester

  • MDL number MFCD08062950
  • Cas Number : 178876-82-9
  • Molecular Weight : 231.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin GB
Product Link https://www.manchesterorganics.com/K23003
Availability details for
Quantity
  • Availability
  • Add to Cart