MAN326368384 Manchester Organics

6-Chlorosalicylic acid

  • MDL number MFCD10687031
  • Molecular Weight : 172.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.94
Country of Origin GB
Product Link https://www.manchesterorganics.com/K23074
Availability details for
Quantity
  • Availability
  • Add to Cart