MAN326368390 Manchester Organics

4-Bromo-2-pyridinemethanol

  • MDL number MFCD09800557
  • Cas Number : 131747-45-0
  • Molecular Weight : 188.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/R23079
Availability details for
Quantity
  • Availability
  • Add to Cart