MAN326368396 Manchester Organics

3-Chloro-2-nitrobenzotrifluoride

  • MDL number MFCD00272563
  • Molecular Weight : 225.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/W22445

Safety Information

MSDS https://www.manchesterorganics.com/W22445
Availability details for
Quantity
  • Availability
  • Add to Cart