MAN326368405 Manchester Organics

2-Chloro-3-fluoro-4-iodopyridine

  • MDL number MFCD04972389
  • Cas Number : 148639-07-0
  • Molecular Weight : 257.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin GB
Product Link https://www.manchesterorganics.com/Y23086
Availability details for
Quantity
  • Availability
  • Add to Cart