MAN326368437 Manchester Organics

5-Amino-3-fluorobenzoic acid

  • MDL number MFCD07779526
  • Molecular Weight : 155.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/F13011
Availability details for
Quantity
  • Availability
  • Add to Cart