MAN326368478 Manchester Organics

4'-[(Trifluoromethylthio]-1,1'-biphenyl-2-amine

  • MDL number MFCD08447253
  • Molecular Weight : 269.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link https://www.manchesterorganics.com/A13052

Safety Information

MSDS https://www.manchesterorganics.com/A13052
Availability details for
Quantity
  • Availability
  • Add to Cart